3-Bromo-N-cyclohexyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBCA-4380-5GEA 0 GBP
COBBCA-4380-5G COBBCA-4380-1G
3-Bromo-N-cyclohexyl-5-nitrobenzamide
3-Bromo-N-cyclohexyl-5-nitrobenzamide
Formula: C₁₃H₁₅BrN₂O₃
MDL Number: MFCD09475857
CAS Number: 941294-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR