4-Bromo-N-cyclopentylbenzamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4411-1GEA
39.57
GBP
COBBCA-4411-1G
COBBCA-4411-5G
4-Bromo-N-cyclopentylbenzamide
4-Bromo-N-cyclopentylbenzamide
Formula:
C₁₂H₁₄BrNO |
MDL Number:
MFCD00588998 CAS Number: 223557-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...