4-Bromo-N-cyclohexylbenzamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4424-25GEA
0
GBP
COBBCA-4424-25G
COBBCA-4424-1G
COBBCA-4424-5G
4-Bromo-N-cyclohexylbenzamide
4-Bromo-N-cyclohexylbenzamide
Formula:
C₁₃H₁₆BrNO |
MDL Number:
MFCD00451750 CAS Number: 223553-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...