3-Bromo-N-cyclohexylbenzamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4476-5GEA
0
GBP
COBBCA-4476-5G
COBBCA-4476-1G
3-Bromo-N-cyclohexylbenzamide
3-Bromo-N-cyclohexylbenzamide
Formula:
C₁₃H₁₆BrNO |
MDL Number:
MFCD00554081 CAS Number: 59507-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...