Potassium-2-hydroxyphenyltrifluoroborate
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Potassium trifluoro(2-hydroxyphenyl)borate
COBBTB-0019-5GEA
0
GBP
COBBTB-0019-5G
COBBTB-0019-1G
Potassium-2-hydroxyphenyltrifluoroborate
Potassium 2-hydroxyphenyltrifluoroborate
Formula:
C₆H₅BF₃KO MW: 200.0078 g/mol |
MDL Number:
MFCD07781243 CAS Number: 850313-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...