3-Amino-3-(4-fluorophenyl)propionic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL573418-1GEA
46.3
GBP
SIAL573418-1G
3-Amino-3-(4-fluorophenyl)propionic acid, Sigma-Aldrich®
3-Amino-3-(4-fluorophenyl)propionic acid
Formula:
C₉H₁₀FNO₂ MW: 183.18 g/mol |
MDL Number:
MFCD00181811 CAS Number: 325-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...