Potassium benzyltrifluoroborate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL563056-1GEA
150
GBP
SIAL563056-1G
SIAL563056-5G
Potassium benzyltrifluoroborate, Sigma-Aldrich®
Potassium benzyltrifluoroborate
Formula:
C₇H₇BF₃K MW: 198.0350 g/mol |
MDL Number:
MFCD03427261 CAS Number: 329976-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...