Isopropylboronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
, Isopropylboronic acid Pinacol ester
, 2-Isopropylboronic acid, pinacol ester
COBBPN-8639-5GEA
0
GBP
COBBPN-8639-5G
COBBPN-8639-1G
Isopropylboronic acid pinacol ester
Isopropylboronic acid pinacol ester
Formula:
C₉H₁₉BO₂ MW: 170.06 g/mol |
MDL Number:
MFCD05663856 CAS Number: 76347-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...