Iodomethyl methyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Stabilisation:
Stabilised with copper
SIAL380229-5GEA
85.6
GBP
SIAL380229-5G
Iodomethyl methyl ether stabilised, Sigma-Aldrich®
Iodomethyl methyl ether
Formula:
C₂H₅IO |
MDL Number:
MFCD00134516 CAS Number: 13057-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...