6-Acetamido-2,3-difluoroanisole
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBAN-1362-5GEA
0
GBP
COBBAN-1362-5G
COBBAN-1362-1G
6-Acetamido-2,3-difluoroanisole
6-Acetamido-2,3-difluoroanisole
Formula:
C₉H₉F₂NO₂ |
MDL Number:
MFCD11053863 CAS Number: 1065073-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...