2',4'-Dichloro-5'-hydroxyacetanilide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-(2,4-Dichloro-5-hydroxyphenyl)acetamide
M-Clarity™ quality level = MQ100
SIAL655880-5GEA
426
GBP
SIAL655880-5G
SIAL655880-1G
2',4'-Dichloro-5'-hydroxyacetanilide, Sigma-Aldrich®
2',4'-Dichloro-5'-hydroxyacetanilide
Formula:
C₈H₇Cl₂NO₂ MW: 220.0527 g/mol |
MDL Number:
MFCD00186236 CAS Number: 67669-19-6 EINECS: 429-110-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...