Dimethachlor
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC12670000EA
182
GBP
EHERC12670000
Dimethachlor
Dimethachlor
Formula:
C₁₃H₁₈ClNO₂ |
MDL Number:
MFCD00055472 CAS Number: 50563-36-5 EINECS: 256-625-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...