Trimethylsilyl bromoacetate, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SAFA235113-25GEA 117 GBP
SAFA235113-25G
Trimethylsilyl bromoacetate, Sigma-Aldrich®
Trimethylsilyl bromoacetate
Formula: C₅H₁₁BrO₂Si
MW: 211,13
MDL Number: MFCD00000187
CAS Number: 18291-80-0
EINECS: 242-165-3
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR