DL-Penicillamine
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE13853922-25GEA
0
GBP
MOLE13853922-25G
DL-Penicillamine
DL-Penicillamine
Formula:
C₅H₁₁NO₂S MW: 149,21 |
MDL Number:
MFCD00004856 CAS Number: 52-66-4 EINECS: 200-147-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...