1,2:7,8-Diepoxyoctane for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1,4-Di(oxiran-2-yl)butane
M-Clarity™ quality level = MQ200
8.21770.0010EA
133
GBP
8.21770.0010
8.21770.9999
1,2:7,8-Diepoxyoctane for synthesis, Sigma-Aldrich®
1,2:7,8-Diepoxyoctane
Formula:
C₈H₁₄O₂ MW: 142.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005155 CAS Number: 2426-07-5 EINECS: 219-375-9 UN: 2922 ADR: 8 (6.1),II |
Specification Test Results
Solubility | 7 g/l (20 °C) |
Melting Point | 7 °C |
Molar Mass | 142.19 g/mol |
Boiling Point | 222 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 89 °C DIN 51758 |
Refractive Index | 1.4456 (20 °C, 589 nm) |
Density | 0.98 g/cm3 (20 °C) |
Ignition Point | 350 °C DIN 51794 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...