1,2:3,4-Diepoxybutane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Butadiene diepoxide
, 1,3-Butadiene diepoxide
M-Clarity™ quality level = MQ200
SIAL202533-1GEA
70.8
GBP
SIAL202533-1G
SIAL202533-5G
SIAL202533-25G
1,2:3,4-Diepoxybutane, Sigma-Aldrich®
1,2:3,4-Diepoxybutane
Formula:
C₄H₆O₂ MW: 86.09 g/mol |
MDL Number:
MFCD00005120 CAS Number: 1464-53-5 EINECS: 215-979-1 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...