Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2:3,4-Diepoxybutane ≥98%, AcroSeal®

Supplier: Acros Organics
Danger
Synonyms: 1,3-Butadiene diepoxideButadiene diepoxide



Formula: C₄H₆O₂
MW: 86,09
Boiling Pt: 56…58 °C (25 mmHg)
Melting Pt: 2…4 °C
Density: 1,11 g/cm³
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005120
CAS Number: 1464-53-5
EINECS: 215-979-1
UN: 3489
ADR: 6.1,I
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Authentic
GC <=96.0%
Refractive index 1.4325 to 1.4345 (20°C, 589 nm)

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
AcroSeal glass bottle 50 ml ACRO330070500
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Density 1.11
Flash point 45°C

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty