2-Acetamidothiazole ≥98%

Supplier: Alfa Aesar
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B23873.06EA 23.6 GBP
B23873.06 B23873.14
2-Acetamidothiazole ≥98%
2-Acetamidothiazole
Formula: C₅H₆N₂OS
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00016892
CAS Number: 2719-23-5
EINECS: 220-321-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR