2,4-Dichloro-5-cyanothiazole
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHI-1211-1GEA
39.57
GBP
COBBHI-1211-1G
COBBHI-1211-250M
2,4-Dichloro-5-cyanothiazole
2,4-Dichloro-5-cyanothiazole
Formula:
C₄Cl₂N₂S MW: 179.0300 g/mol |
MDL Number:
MFCD08669569 CAS Number: 82554-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...