Sodium borodeuteride (99% D), rods
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
35102.06EA
176
GBP
35102.06
35102.03
Sodium borodeuteride (99% D), rods
Sodium borodeuteride
Formula:
D₄BNa MW: 41.857 g/mol Melting Pt: ∼400 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003519 CAS Number: 15681-89-7 EINECS: 239-764-7 UN: 1426 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...