1,4-Diamino-2,3-dihydroanthraquinone for synthesis
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,4-Diamino-2,3-dihydroanthracene-9,10-dione
M-Clarity™ quality level = MQ200
8.43852.0100EA
150
GBP
8.43852.0100
8.43852.0010
1,4-Diamino-2,3-dihydroanthraquinone for synthesis
1,4-Diamino-2,3-dihydroanthraquinone
Formula:
C₁₄H₁₂N₂O₂ MW: 240.2600 g/mol |
MDL Number:
MFCD00035689 CAS Number: 81-63-0 EINECS: 201-367-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...