2-Benzylamino-5-bromopyrimidine

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Benzyl-5-bromopyrimidin-2-amine

COBBPY-8009-5GEA 0 GBP
COBBPY-8009-5G COBBPY-8009-1G
2-Benzylamino-5-bromopyrimidine
2-Benzylamino-5-bromopyrimidine
Formula: C₁₁H₁₀BrN₃
MDL Number: MFCD00483264
CAS Number: 38373-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR