5-Bromo-4-(4-ethylphenyl)pyrimidine

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBHC-6082-1GEA 31.66 GBP
COBBHC-6082-1G COBBHC-6082-5G
5-Bromo-4-(4-ethylphenyl)pyrimidine
5-Bromo-4-(4-ethylphenyl)pyrimidine
Formula: C₁₂H₁₁BrN₂
MDL Number: MFCD09800937
CAS Number: 951884-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR