1H-Pyrazole-3-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Pyrazolecarboxylic acid

M-Clarity™ quality level = MQ100

SIAL707384-1GEA 37.2 GBP
SIAL707384-1G
1H-Pyrazole-3-carboxylic acid, Sigma-Aldrich®
1H-Pyrazole-3-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
MDL Number: MFCD00077436
CAS Number: 1621-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR