Methyl-2-pyrazinecarboxylate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl pyrazine-2-carboxylate
, Methyl 2-pyrazinecarboxylate
M-Clarity™ quality level = MQ100
SIAL519677-5GEA
199
GBP
SIAL519677-5G
Methyl-2-pyrazinecarboxylate, Sigma-Aldrich®
Methyl 2-pyrazinecarboxylate
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00014611 CAS Number: 6164-79-0 EINECS: 228-198-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...