Chlorobis(4-methoxyphenyl)phosphine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL710377-2GEA
228
GBP
SIAL710377-2G
Chlorobis(4-methoxyphenyl)phosphine, Sigma-Aldrich®
Chlorobis(4-methoxyphenyl)phosphine
Formula:
C₁₄H₁₄ClO₂P MW: 280.69 g/mol |
MDL Number:
MFCD04972301 CAS Number: 13685-30-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...