Dicyclohexylchlorophosphine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Chlorodicyclohexylphosphine
M-Clarity™ quality level = MQ100
SIAL481408-1GEA
115
GBP
SIAL481408-1G
SIAL481408-5G
Dicyclohexylchlorophosphine, Sigma-Aldrich®
Dicyclohexylchlorophosphine
Formula:
C₁₂H₂₂ClP MW: 232.73 g/mol |
MDL Number:
MFCD00014294 CAS Number: 16523-54-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...