Allyl cyclohexanepropionate ≥98% FCC, FG, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Allyl 3-cyclohexylpropionateCyclohexanepropionic acid allyl esterAllyl 3-cyclohexylpropanoateCyclohexanepropanoic acid 2-propen-1-yl ester2-Propenyl 3-cyclohexylpropanoate

M-Clarity™ quality level = MQ300

SIALW202606-10KG-KEA 599 GBP
SIALW202606-10KG-K SIALW202606-1KG-K SIALW202606-5KG-K
Allyl cyclohexanepropionate ≥98% FCC, FG, Sigma-Aldrich®
Allyl cyclohexanepropionate
Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
MDL Number: MFCD00059557
CAS Number: 2705-87-5
EINECS: 220-292-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR