Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Samarium diiodide 1 M in THF stabilised, AcroSeal®

Supplier: Acros Organics
Danger
Synonyms: Samarium (II) iodide

Stabilisation: Stabilised with Sm

ACRO294991000
Samarium diiodide 1 M in THF stabilised, AcroSeal®


Formula: SmI₂
MW: 404,17 g/mol
Density: 0,922
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058873
CAS Number: 32248-43-4
EINECS: 000-000-0
UN: 2056
ADR: 3,II
Order Now
Specification Test Results
Appearance Blue-green to dark blue Solution
Titration Argentometric 0.08 to 0.12 M
Stabilizer Samarium

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty