Bis(trimethylsilyl)methane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL282138-5GEA
463
GBP
SIAL282138-5G
Bis(trimethylsilyl)methane, Sigma-Aldrich®
Bis(trimethylsilyl)methane
Formula:
C₇H₂₀Si₂ MW: 160.405 g/mol |
MDL Number:
MFCD00008275 CAS Number: 2117-28-4 EINECS: 218-322-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...