3-(4-Boc-piperazino)-4-nitroanisole

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBHC-3091-5GEA 0 GBP
COBBHC-3091-5G COBBHC-3091-1G
3-(4-Boc-piperazino)-4-nitroanisole
3-(4-Boc-piperazino)-4-nitroanisole
Formula: C₁₆H₂₃N₃O₅
MDL Number: MFCD13195696
CAS Number: 1215205-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR