6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBPY-7141-1GEA
0
GBP
COBBPY-7141-1G
6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridine
Formula:
C₇H₆BrN₃S |
MDL Number:
MFCD09475863 CAS Number: 941294-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...