2-Amino-4-(4-pyridyl)thiazole
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(Pyridin-4-yl)thiazol-2-amine
CHMPCFF406.1EA
0
GBP
CHMPCFF406.1
CHMPCFF406.2.5
CHMPCFF406.5
FLUO017429.1G
FLUO017429.5G
FLUO017429-2.5G
2-Amino-4-(4-pyridyl)thiazole
2-Amino-4-(4-pyridyl)thiazole
Formula:
C₈H₇N₃S MW: 177.22 g/mol |
MDL Number:
MFCD00490009 CAS Number: 30235-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...