2-(4-Pyridinyl)-4-thiazolecarboxylic acid
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAYBCC34701DAEA
0
GBP
MAYBCC34701DA
2-(4-Pyridinyl)-4-thiazolecarboxylic acid
2-(4-Pyridinyl)-4-thiazolecarboxylic acid
Formula:
C₉H₆N₂O₂S MW: 206,22 |
MDL Number:
MFCD00171745 CAS Number: 21278-86-4 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...