4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid
M-Clarity™ quality level = MQ100
SIAL702536-1GEA
139
GBP
SIAL702536-1G
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid, Sigma-Aldrich®
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid
Formula:
C₁₀H₈N₂O₂S MW: 220.2477 g/mol |
MDL Number:
MFCD00171757 CAS Number: 144060-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...