Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate
H54942.06EA
169
GBP
H54942.06
Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate ≥97%
Ethyl 4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate
Formula:
C₁₂H₁₂N₂O₂S MW: 248.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01567252 CAS Number: 39091-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...