7-Oxabicyclo[2.2.1]heptane, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SAFAO8704-5GEA 107 GBP
SAFAO8704-5G
7-Oxabicyclo[2.2.1]heptane, Sigma-Aldrich®
7-Oxabicyclo[2.2.1]heptane
Formula: C₆H₁₀O
MW: 98,15
MDL Number: MFCD00082342
CAS Number: 279-49-2
EINECS: 205-998-3
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR