3-Nitro-1,8-naphthalic anhydride, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SAFAN19001-25GEA 0 GBP
SAFAN19001-25G SAFAN19001-5G
3-Nitro-1,8-naphthalic anhydride, Sigma-Aldrich®
3-Nitro-1,8-naphthalic anhydride
Formula: C₁₂H₅NO₅
MDL Number: MFCD00006926
CAS Number: 3027-38-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR