HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Supplier: Enzo Life Sciences
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
, HBTU
ALEXALX121003G250EA
996
GBP
ALEXALX121003G250
ALEXALX121003G100
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Formula:
C₁₁H₁₆F₆N₅OP MW: 379.2 g/mol |
MDL Number:
MFCD00075445 CAS Number: 94790-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...