TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
TBTU
BACE4013268.0025EA
61.31
GBP
BACE4013268.0025
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula:
C₁₁H₁₆BF₄N₅O MW: 321.1 g/mol |
MDL Number:
MFCD00077413 CAS Number: 125700-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...