O-Methylisourea hemisulphate, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: O-Methylisouronium sulphate

SAFA455598-100GEA 263 GBP
SAFA455598-100G
O-Methylisourea hemisulphate, Sigma-Aldrich®
O-Methylisourea hemisulphate
Formula: C₂H₆N₂O·0.5H₂SO₄
MW: 123,12 g/mol
MDL Number: MFCD00040594
CAS Number: 52328-05-9
EINECS: 257-851-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR