1-(2-Furoyl)piperazine, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SAFA558966-25GEA 76.4 GBP
SAFA558966-25G
1-(2-Furoyl)piperazine, Sigma-Aldrich®
1-(2-Furoyl)piperazine
Formula: C₉H₁₂N₂O₂
MW: 180,21
MDL Number: MFCD00038831
CAS Number: 40172-95-0
EINECS: 254-823-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR