Iminostilbene
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE12960416-100GEA
0
GBP
MOLE12960416-100G
Iminostilbene
Iminostilbene
Formula:
C₁₄H₁₁N MW: 193,25 |
MDL Number:
MFCD00005071 CAS Number: 256-96-2 EINECS: 205-970-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...