Iminostilbene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC14285500EA 101 GBP
EHERC14285500
Iminostilbene
Iminostilbene
Formula: C₁₄H₁₁N
MW: 193,25
MDL Number: MFCD00005071
CAS Number: 256-96-2
EINECS: 205-970-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR