1-(2-Tetrahydrofuroyl)piperazine hydrobromide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Piperazin-1-yl(tetrahydrofuran-2-yl)methanone hydrobromide
M-Clarity™ quality level = MQ100
SIAL570990-5GEA
108
GBP
SIAL570990-5G
1-(2-Tetrahydrofuroyl)piperazine hydrobromide, Sigma-Aldrich®
1-(2-Tetrahydrofuroyl)piperazine hydrobromide
Formula:
C₉H₁₇BrN₂O₂ MW: 265.1475 g/mol |
MDL Number:
MFCD03701619 CAS Number: 63590-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...