9-Phenylcarbazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIALP21501-10GEA
129
GBP
SIALP21501-10G
9-Phenylcarbazole, Sigma-Aldrich®
9-Phenylcarbazole
Formula:
C₁₈H₁₃N MW: 243.30 g/mol |
MDL Number:
MFCD00004965 CAS Number: 1150-62-5 EINECS: 214-564-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...