2,1,3-Benzothiadiazol-4-ylamine, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Aminobenzo-2,1,3-thiadiazole

SAFA102520-1GEA 89.1 GBP
SAFA102520-1G
2,1,3-Benzothiadiazol-4-ylamine, Sigma-Aldrich®
2,1,3-Benzothiadiazol-4-ylamine
Formula: C₆H₅N₃S
MW: 151,19
MDL Number: MFCD00005810
CAS Number: 767-64-6
EINECS: 212-186-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR