Estrone

Supplier: U.S. Pharmacopeia
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USPH1255001EA 310 GBP
USPH1255001
Estrone
Estrone
Formula: C₁₈H₂₂O₂
MDL Number: MFCD00003620
CAS Number: 53-16-7
EINECS: 200-164-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR