Estrone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
SIALE9750-1GEA
79.3
GBP
SIALE9750-1G
SIALE9750-5G
SIALE9750-500MG
SIALE9750-25G
Estrone, Sigma-Aldrich®
Estrone
Formula:
C₁₈H₂₂O₂ |
MDL Number:
MFCD00003620 CAS Number: 53-16-7 EINECS: 200-164-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...