Estrone, Supelco®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

46573-250MGEA 28.4 GBP
46573-250MG
Estrone, Supelco®
Estrone
Formula: C₁₈H₂₂O₂
MDL Number: MFCD00003620
CAS Number: 53-16-7
EINECS: 200-164-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR